trans-2-Fluorocyclohexanol ≥98.0%

Supplier: TCI America

F0631-1G F0631-5G
TCF0631-1GEA 180.12 CAD
TCF0631-1G TCF0631-5G
trans-2-Fluorocyclohexanol ≥98.0%
trans-2-Fluorocyclohexanol

CAS Number: 656-60-0
MDL Number: MFCD08146648
Molecular Formula: C6H11FO
Molecular Weight: 118.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 166
Melting point (°C): 21
Storage Temperature: 0-10°C

Formula: C₆H₁₁FO
Boiling Pt: 166 °C
Melting Pt: 21 °C
MDL Number: MFCD08146648
CAS Number: 656-60-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a22e73d5a3d74829c2c6c179939f5a5aa60bb967&vid=a22e73d5a3d74829c2c6c179939f5a5aa60bb967&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR