3-Fluorosalicylaldehyde ≥98.0%

Supplier: TCI America

Synonyms: 3-Fluoro-2-hydroxybenzaldehyde

F0625-1G F0625-5G
TCF0625-1GEA 112.18 CAD
TCF0625-1G TCF0625-5G
3-Fluorosalicylaldehyde ≥98.0%
3-Fluorosalicylaldehyde

CAS Number: 394-50-3
MDL Number: MFCD00003319
Molecular Formula: C7H5FO2
Molecular Weight: 140.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 70


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Fluoro-2-hydroxybenzaldehyde

Formula: C₇H₅FO₂
MW: 140.12 g/mol
Melting Pt: 70 °C
MDL Number: MFCD00003319
CAS Number: 394-50-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d27509177559cf965055f8b771a89319b46c071&vid=1d27509177559cf965055f8b771a89319b46c071&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR