α,α,α,6-Tetrafluoro-m-toluic acid ≥98.0%

Supplier: TCI America

Synonyms: 2-Fluoro-5-(trifluoromethyl)benzoic acid

F0620-5G F0620-1G
TCF0620-5GEA 132.25 CAD
TCF0620-5G TCF0620-1G
α,α,α,6-Tetrafluoro-m-toluic acid ≥98.0%
α,α,α,6-Tetrafluoro-m-toluic acid

CAS Number: 115029-23-7
MDL Number: MFCD00061292
Molecular Formula: C8H4F4O2
Molecular Weight: 208.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 100


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha,alpha,alpha,6-Tetrafluoro-m-toluic Acid

Formula: C₈H₄F₄O₂
MW: 208.11 g/mol
Melting Pt: 100 °C
MDL Number: MFCD00061292
CAS Number: 115029-23-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d147df97cf220f5c8057cc4a1a1f40d4d70f5464&vid=d147df97cf220f5c8057cc4a1a1f40d4d70f5464&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR