α,α,α,2-Tetrafluoro-m-toluic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

F0619-5G F0619-1G
TCF0619-5GEA 124.82 CAD
TCF0619-5G TCF0619-1G
α,α,α,2-Tetrafluoro-m-toluic acid ≥98.0% (by GC, titration analysis)
α,α,α,2-Tetrafluoro-m-toluic acid

CAS Number: 115029-22-6
MDL Number: MFCD00040980
Molecular Formula: C8H4F4O2
Molecular Weight: 208.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 127


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha,alpha,alpha,2-Tetrafluoro-m-toluic Acid

Formula: C₈H₄F₄O₂
MW: 208.11 g/mol
Melting Pt: 127 °C
MDL Number: MFCD00040980
CAS Number: 115029-22-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2b0482699a2e58804c21e9d16b871ac887a05b3&vid=e2b0482699a2e58804c21e9d16b871ac887a05b3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR