5-Formyl-2-furanylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
F0611-1G
F0611-5G
TCF0611-1GEA
33.18
CAD
TCF0611-1G
TCF0611-5G
5-Formyl-2-furanylboronic acid (contains varying amounts of Anhydride)
5-Formyl-2-furanylboronic acid
CAS Number: 27329-70-0
MDL Number: MFCD01114696
Molecular Formula: C5H5BO4
Molecular Weight: 139.90
Form: Crystal
Color: Pale Yellow
Melting point (°C): 151
Formula:
C₅H₅BO₄ Melting Pt: 151 °C |
MDL Number:
MFCD01114696 CAS Number: 27329-70-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...