N(α)-Fmoc-D-tryptophan ≥97.0%

Supplier: TCI America

Synonyms: Fmoc-D-Trp-OH

F0609-5G F0609-25G
TCF0609-5GEA 93.22 CAD
TCF0609-5G TCF0609-25G
N(α)-Fmoc-D-tryptophan ≥97.0%
N(α)-Fmoc-D-tryptophan

CAS Number: 86123-11-7
MDL Number: MFCD00062954
Molecular Formula: C26H22N2O4
Molecular Weight: 426.47
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 184
Specific rotation [a]20/D: 29 deg (C=1, DMF)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Fmoc-D-tryptophan, Fmoc-D-Trp-OH

Formula: C₂₆H₂₂N₂O₄
MW: 426.46 g/mol
Melting Pt: 184 °C
MDL Number: MFCD00062954
CAS Number: 86123-11-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad6ac901b7206eceb28c14ec8f0245f325feabac&vid=ad6ac901b7206eceb28c14ec8f0245f325feabac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR