Fmoc-D-phenylalanine ≥98.0%

Supplier: TCI America

Synonyms: Fmoc-D-Phe-OH, N-Fmoc-D-phenylalanine

F0605-5G F0605-25G
TCF0605-5GEA 58.46 CAD
TCF0605-5G TCF0605-25G
Fmoc-D-phenylalanine ≥98.0%
Fmoc-D-phenylalanine

CAS Number: 86123-10-6
MDL Number: MFCD00062955
Molecular Formula: C24H21NO4
Molecular Weight: 387.44
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 183
Specific rotation [a]20/D: 38 deg (C=1, DMF)


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Fmoc-D-phenylalanine, Fmoc-D-Phe-OH

Formula: C₂₄H₂₁NO₄
Melting Pt: 183 °C
MDL Number: MFCD00062955
CAS Number: 86123-10-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe1084c6aa269b388724e82f8920d9c06a3ae126&vid=fe1084c6aa269b388724e82f8920d9c06a3ae126&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR