9-Fluorenone-1-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
F0590-1G
F0590-5G
TCF0590-1GEA
80.58
CAD
TCF0590-1G
TCF0590-5G
9-Fluorenone-1-carboxylic acid ≥98.0% (by HPLC, titration analysis)
9-Fluorenone-1-carboxylic acid
CAS Number: 1573-92-8
MDL Number: MFCD00011537
Molecular Formula: C14H8O3
Molecular Weight: 224.22
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 198
Formula:
C₁₄H₈O₃ Melting Pt: 198 °C |
MDL Number:
MFCD00011537 CAS Number: 1573-92-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...