5-Fluoroisatin ≥98.0% (by GC, titration analysis)

Supplier: TCI America

F0589-25G F0589-5G
TCF0589-25GEA 178.54 CAD
TCF0589-25G TCF0589-5G
5-Fluoroisatin ≥98.0% (by GC, titration analysis)
5-Fluoroisatin

CAS Number: 443-69-6
MDL Number: MFCD00022795
Molecular Formula: C8H4FNO2
Molecular Weight: 165.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 227


Product Categories: Synthetic Organic Chemistry, Isatins (Multi-component Reaction), Fluorinated Building Blocks
Other Product Names / Synonyms: 5-Fluoroindole-2,3-dione, 5-Fluoro-2,3-indolinedione

Formula: C₈H₄FNO₂
Melting Pt: 227 °C
MDL Number: MFCD00022795
CAS Number: 443-69-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fa8b357345e76ecbe85eb6436f4bd73a713952d&vid=5fa8b357345e76ecbe85eb6436f4bd73a713952d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR