α,α,α,4-Tetrafluoro-o-toluic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

F0577-5G F0577-1G
TCF0577-5GEA 153.26 CAD
TCF0577-5G TCF0577-1G
α,α,α,4-Tetrafluoro-o-toluic acid ≥97.0% (by GC, titration analysis)
α,α,α,4-Tetrafluoro-o-toluic acid

CAS Number: 141179-72-8
MDL Number: MFCD00040982
Molecular Formula: C8H4F4O2
Molecular Weight: 208.11
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 123


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha,alpha,alpha,4-Tetrafluoro-o-toluic Acid

Formula: C₈H₄F₄O₂
Melting Pt: 123 °C
MDL Number: MFCD00040982
CAS Number: 141179-72-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c83f603dec420683fd79935d290a45b2a7845341&vid=c83f603dec420683fd79935d290a45b2a7845341&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR