5-Fluorosalicylaldehyde ≥98.0% (by GC, titration analysis)

Supplier: TCI America

F0574-25G F0574-5G
TCF0574-25GEA 327.06 CAD
TCF0574-25G TCF0574-5G
5-Fluorosalicylaldehyde ≥98.0% (by GC, titration analysis)
5-Fluorosalicylaldehyde

CAS Number: 347-54-6
MDL Number: MFCD01090997
Molecular Formula: C7H5FO2
Molecular Weight: 140.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 56
Melting point (°C): 85


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 5-Fluoro-2-hydroxybenzaldehyde

Formula: C₇H₅FO₂
Boiling Pt: 56 °C
Melting Pt: 85 °C
MDL Number: MFCD01090997
CAS Number: 347-54-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0ed9e06d18b8e9ed9fdc945ddaa40cf109f98b1&vid=c0ed9e06d18b8e9ed9fdc945ddaa40cf109f98b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR