2-Fluoro-4-methoxybenzoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

F0572-25G F0572-5G
TCF0572-25GEA 383.94 CAD
TCF0572-25G TCF0572-5G
2-Fluoro-4-methoxybenzoic acid ≥98.0% (by titrimetric analysis)
2-Fluoro-4-methoxybenzoic acid

CAS Number: 394-42-3
MDL Number: MFCD00509373
Molecular Formula: C8H7FO3
Molecular Weight: 170.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 193


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Fluoro-p-anisic Acid

Formula: C₈H₇FO₃
Melting Pt: 193 °C
MDL Number: MFCD00509373
CAS Number: 394-42-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78418d5892ed60e0da88bec5d173374d87c10f2b&vid=78418d5892ed60e0da88bec5d173374d87c10f2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR