3-Fluoro-4-nitrotoluene ≥98.0%

Supplier: TCI America

F0567-25G F0567-5G
TCF0567-25GEA 323.9 CAD
TCF0567-25G TCF0567-5G
3-Fluoro-4-nitrotoluene ≥98.0%
3-Fluoro-4-nitrotoluene

CAS Number: 446-34-4
MDL Number: MFCD00007053
Molecular Formula: C7H6FNO2
Molecular Weight: 155.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 98
Melting point (°C): 54
Flash Point (°C): 110

Formula: C₇H₆FNO₂
Boiling Pt: 98 °C
Melting Pt: 54 °C
Flash Pt: 110 °C
MDL Number: MFCD00007053
CAS Number: 446-34-4
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8414b5d277280ff6a15ff087310c883d85e0d016&vid=e0e87a029b8e38b629495cb24d8dc539a4a8feb0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR