4-Fluoro-o-toluic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

F0559-25G F0559-5G
TCF0559-25GEA 598.35 CAD
TCF0559-25G TCF0559-5G
4-Fluoro-o-toluic acid ≥98.0% (by GC, titration analysis)
4-Fluoro-o-toluic acid

CAS Number: 321-21-1
MDL Number: MFCD04115880
Molecular Formula: C8H7FO2
Molecular Weight: 154.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 174


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Fluoro-o-toluic Acid

Formula: C₈H₇FO₂
Melting Pt: 174 °C
MDL Number: MFCD04115880
CAS Number: 321-21-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89673faa559ffb170a7bd4232258e53f46fd3fa0&vid=89673faa559ffb170a7bd4232258e53f46fd3fa0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR