3-Fluoro-p-toluic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

F0552-25G F0552-5G
TCF0552-25GEA 361.82 CAD
TCF0552-25G TCF0552-5G
3-Fluoro-p-toluic acid ≥98.0% (by GC, titration analysis)
3-Fluoro-p-toluic acid

CAS Number: 350-28-7
MDL Number: MFCD00002490
Molecular Formula: C8H7FO2
Molecular Weight: 154.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 173


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Fluoro-p-toluic Acid

Formula: C₈H₇FO₂
Melting Pt: 173 °C
MDL Number: MFCD00002490
CAS Number: 350-28-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9918b77148bb01c56116242d08156e8287e7322c&vid=9918b77148bb01c56116242d08156e8287e7322c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR