7-Fluoroisatin ≥98.0% (by GC, titration analysis)
Supplier: TCI America
F0551-5G
F0551-25G
TCF0551-5GEA
137.14
CAD
TCF0551-5G
TCF0551-25G
7-Fluoroisatin ≥98.0% (by GC, titration analysis)
7-Fluoroisatin
CAS Number: 317-20-4
Molecular Formula: C8H4FNO2
Molecular Weight: 165.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 195
Product Categories: Synthetic Organic Chemistry, Isatins (Multi-component Reaction), Fluorinated Building Blocks
Other Product Names / Synonyms: 7-Fluoroindole-2,3-dione, 7-Fluoro-2,3-indolinedione
Formula:
C₈H₄FNO₂ Melting Pt: 195 °C |
MDL Number:
MFCD01569508 CAS Number: 317-20-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...