5-Formyl-2-thiopheneboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

F0549-5G F0549-1G
TCF0549-5GEA 170.64 CAD
TCF0549-5G TCF0549-1G
5-Formyl-2-thiopheneboronic acid (contains varying amounts of Anhydride)
5-Formyl-2-thiopheneboronic acid

CAS Number: 4347-33-5
Molecular Formula: C5H5BO3S
Molecular Weight: 155.96
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 135
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 5-Formyl-2-thienylboronic Acid

Formula: C₅H₅BO₃S
Melting Pt: 135 °C
MDL Number: MFCD02093666
CAS Number: 4347-33-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=953259747aa6e2eb98fb0a5c55cc7df1fd3a509b&vid=953259747aa6e2eb98fb0a5c55cc7df1fd3a509b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR