4-Fluoro-3-(trifluoromethyl)benzaldehyde ≥97.0%
Supplier: TCI America
F0545-5G
F0545-1G
F0545-25G
TCF0545-5GEA
157.05
CAD
TCF0545-5G
TCF0545-1G
TCF0545-25G
4-Fluoro-3-(trifluoromethyl)benzaldehyde ≥97.0%
4-Fluoro-3-(trifluoromethyl)benzaldehyde
CAS Number: 67515-60-0
Molecular Formula: C8H4F4O
Molecular Weight: 192.11
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 60
Flash Point (°C): 80
Specific Gravity (20/20): 1.41
Formula:
C₈H₄F₄O MW: 192.11 g/mol Boiling Pt: 60 °C Density: 1.41 (20 °C) Flash Pt: 80 °C |
MDL Number:
MFCD00061314 CAS Number: 67515-60-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...