3-Fluoro-4-methoxybenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

F0543-1G F0543-5G
TCF0543-1GEA 105.86 CAD
TCF0543-1G TCF0543-5G
3-Fluoro-4-methoxybenzoic acid ≥98.0% (by GC, titration analysis)
3-Fluoro-4-methoxybenzoic acid

CAS Number: 403-20-3
MDL Number: MFCD00060675
Molecular Formula: C8H7FO3
Molecular Weight: 170.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 212


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Fluoro-p-anisic Acid

Formula: C₈H₇FO₃
Melting Pt: 212 °C
MDL Number: MFCD00060675
CAS Number: 403-20-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73392215eb2b08fd3eb87bda31c009c466292026&vid=73392215eb2b08fd3eb87bda31c009c466292026&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR