Fluorene-1-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

F0541-1G F0541-5G
TCF0541-1GEA 94.8 CAD
TCF0541-1G TCF0541-5G
Fluorene-1-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Fluorene-1-carboxylic acid

CAS Number: 6276-03-5
MDL Number: MFCD00001130
Molecular Formula: C14H10O2
Molecular Weight: 210.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Boiling point (°C): 100
Melting point (°C): 248

Formula: C₁₄H₁₀O₂
Boiling Pt: 100 °C
Melting Pt: 248 °C
MDL Number: MFCD00001130
CAS Number: 6276-03-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=30b35f6ad5b8fd8a45d8e8ae8c252815102599c6&vid=30b35f6ad5b8fd8a45d8e8ae8c252815102599c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR