(4-Fluorophenyl)acetone ≥98.0%
Supplier: TCI America
F0535-5G
F0535-25G
TCF0535-5GEA
42.66
CAD
TCF0535-5G
TCF0535-25G
(4-Fluorophenyl)acetone ≥98.0%
(4-Fluorophenyl)acetone
CAS Number: 459-03-0
MDL Number: MFCD00000362
Molecular Formula: C9H9FO
Molecular Weight: 152.17
Purity/Analysis Method: >98.0% (GC)
Boiling point (°C): 130
Flash Point (°C): 91
Specific Gravity (20/20): 1.11
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-(4-Fluorophenyl)-2-propanone
Formula:
C₉H₉FO Boiling Pt: 130 °C Density: 1.11 (20 °C) Flash Pt: 91 °C |
MDL Number:
MFCD00000362 CAS Number: 459-03-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...