6-Fluoro-m-toluidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

F0528-25G F0528-5G
TCF0528-25GEA 172.22 CAD
TCF0528-25G TCF0528-5G
6-Fluoro-m-toluidine ≥98.0% (by GC, titration analysis)
6-Fluoro-m-toluidine

CAS Number: 452-84-6
Molecular Formula: C7H8FN
Molecular Weight: 125.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 90
Flash Point (°C): 82
Specific Gravity (20/20): 1.11


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Amino-4-fluorotoluene, 6-Fluoro-m-toluidine

Formula: C₇H₈FN
Boiling Pt: 90 °C
Density: 1.11 (20 °C)
Flash Pt: 82 °C
MDL Number: MFCD00007654
CAS Number: 452-84-6
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36ecf54752de90922625d52cc559565670d1c661&vid=36ecf54752de90922625d52cc559565670d1c661&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR