3-Fluoro-o-toluidine ≥98.0%

Supplier: TCI America

Synonyms: 2-Amino-6-fluorotoluene, 3-Fluoro-2-methylaniline

F0526-25G F0526-5G
TCF0526-25GEA 235.42 CAD
TCF0526-25G TCF0526-5G
3-Fluoro-o-toluidine ≥98.0%
3-Fluoro-o-toluidine

CAS Number: 443-86-7
Molecular Formula: C7H8FN
Molecular Weight: 125.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 205
Flash Point (°C): 86
Freezing point (°C): 1
Specific Gravity (20/20): 1.13


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Amino-6-fluorotoluene, 3-Fluoro-o-toluidine

Formula: C₇H₈FN
Boiling Pt: 205 °C
Density: 1.13 (20 °C)
Flash Pt: 86 °C
MDL Number: MFCD00007760
CAS Number: 443-86-7
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c9faf855f101839537d9a80adc8b01b69a6cc35&vid=6c9faf855f101839537d9a80adc8b01b69a6cc35&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR