2-Fluoro-5-iodopyridine ≥98.0%

Supplier: TCI America

F0523-1G F0523-5G
TCF0523-1GEA 61.1 CAD
TCF0523-1G TCF0523-5G
2-Fluoro-5-iodopyridine ≥98.0%
2-Fluoro-5-iodopyridine

CAS Number: 171197-80-1
MDL Number: MFCD03095301
Molecular Formula: C5H3FIN
Molecular Weight: 222.99
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 35
Flash Point (°C): 95

Formula: C₅H₃FIN
Melting Pt: 35 °C
Flash Pt: 95 °C
MDL Number: MFCD03095301
CAS Number: 171197-80-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a88aff3fcff2f4598bdea8809aa59cec19ecd669&vid=a88aff3fcff2f4598bdea8809aa59cec19ecd669&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR