4'-Fluoropropiophenone ≥97.0%

Supplier: TCI America

Synonyms: p-Fluoropropiophenone, 1-(4-Fluorophenyl)propan-1-one

F0522-25G F0522-5G
TCF0522-25GEA 42.66 CAD
TCF0522-25G TCF0522-5G
4'-Fluoropropiophenone ≥97.0%
4'-Fluoropropiophenone

CAS Number: 456-03-1
Molecular Formula: C9H9FO
Molecular Weight: 152.17
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 210
Flash Point (°C): 76
Specific Gravity (20/20): 1.11


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Ethyl 4-Fluorophenyl Ketone

Formula: C₉H₉FO
MW: 152.17
Boiling Pt: 210 °C
Density: 1.09
Flash Pt: 76 °C
MDL Number: MFCD00000356
CAS Number: 456-03-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=465c16fa2cbf6a1841c181202acf741fcc178ef3&vid=465c16fa2cbf6a1841c181202acf741fcc178ef3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR