2-Fluoro-6-methylpyridine ≥95.0%

Supplier: TCI America

Synonyms: 2-Fluoro-6-picoline

F0520-1G F0520-5G
TCF0520-1GEA 33.18 CAD
TCF0520-1G TCF0520-5G
2-Fluoro-6-methylpyridine ≥95.0%
2-Fluoro-6-methylpyridine

CAS Number: 407-22-7
Molecular Formula: C6H6FN
Molecular Weight: 111.12
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 142
Flash Point (°C): 66
Specific Gravity (20/20): 1.08


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 6-Fluoro-2-picoline

Formula: C₆H₆FN
MW: 111.12 g/mol
Boiling Pt: 142 °C
Density: 1.08 (20 °C)
Flash Pt: 66 °C
MDL Number: MFCD00041226
CAS Number: 407-22-7
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04acec02b9963aff6f5e4f684da26e82b82ff1df&vid=04acec02b9963aff6f5e4f684da26e82b82ff1df&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR