4-Fluorobiphenyl ≥98.0%

Supplier: TCI America

F0482-1G F0482-5G
TCF0482-1GEA 112.34 CAD
TCF0482-1G TCF0482-5G
4-Fluorobiphenyl ≥98.0%
4-Fluorobiphenyl

CAS Number: 324-74-3
MDL Number: MFCD00011650
Molecular Formula: C12H9F
Molecular Weight: 172.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 253
Melting point (°C): 75
Flash Point (°C): 110

Formula: C₁₂H₉F
Boiling Pt: 253 °C
Melting Pt: 75 °C
Flash Pt: 110 °C
MDL Number: MFCD00011650
CAS Number: 324-74-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20a0146058ff0a77abfe0b8fa8f43f8551eee421&vid=20a0146058ff0a77abfe0b8fa8f43f8551eee421&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR