3-Fluoro-p-anisaldehyde ≥98.0%

Supplier: TCI America

F0480-1G F0480-5G
TCF0480-1GEA 52.14 CAD
TCF0480-1G TCF0480-5G
3-Fluoro-p-anisaldehyde ≥98.0%
3-Fluoro-p-anisaldehyde

CAS Number: 351-54-2
Molecular Formula: C8H7FO2
Molecular Weight: 154.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 132
Flash Point (°C): 113
Freezing point (°C): 29


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Fluoro-4-methoxybenzaldehyde

Formula: C₈H₇FO₂
Boiling Pt: 132 °C
Flash Pt: 113 °C
MDL Number: MFCD00003349
CAS Number: 351-54-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56956807adbd5a53a33a57ae9da3afd039c8b855&vid=ec84ae7d21209f4bb9ab38aeef0e05e84747ec4e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR