4-(4-Fluorobenzoyl)butyric acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

F0476-1G F0476-5G
TCF0476-1GEA 64.46 CAD
TCF0476-1G TCF0476-5G
4-(4-Fluorobenzoyl)butyric acid ≥98.0% (by GC, titration analysis)
4-(4-Fluorobenzoyl)butyric acid

CAS Number: 149437-76-3
MDL Number: MFCD00667222
Molecular Formula: C11H11FO3
Molecular Weight: 210.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 143


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 5-(4-Fluorophenyl)-5-oxovaleric Acid

Formula: C₁₁H₁₁FO₃
Melting Pt: 143 °C
MDL Number: MFCD00667222
CAS Number: 149437-76-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8e1d1c4cddec094fd9f6804bc0359295ffd652f&vid=a8e1d1c4cddec094fd9f6804bc0359295ffd652f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR