6-Fluoroanthranilic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-Amino-6-fluorobenzoic acid
F0475-1G
F0475-5G
TCF0475-1GEA
42.66
CAD
TCF0475-1G
TCF0475-5G
6-Fluoroanthranilic acid ≥98.0% (by GC, titration analysis)
6-Fluoroanthranilic acid
CAS Number: 434-76-4
MDL Number: MFCD00067781
Molecular Formula: C7H6FNO2
Molecular Weight: 155.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 171
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Amino-6-fluorobenzoic Acid
Formula:
C₇H₆FNO₂ MW: 155.13 g/mol Melting Pt: 171 °C |
MDL Number:
MFCD00067781 CAS Number: 434-76-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...