Methyl (4-fluorobenzoyl)acetate ≥98.0%

Supplier: TCI America

Synonyms: 3-(4-Fluorophenyl)-3-oxopropionic Acid Methyl Ester, Methyl 4-fluorobenzoylacetate, (4-Fluorobenzoyl)acetic Acid Methyl Ester, Methyl 3-(4-Fluorophenyl)-3-oxopropionate

F0472-1G F0472-5G
TCF0472-1GEA 105.86 CAD
TCF0472-1G TCF0472-5G
Methyl (4-fluorobenzoyl)acetate ≥98.0%
Methyl (4-fluorobenzoyl)acetate

CAS Number: 63131-29-3
MDL Number: MFCD00000355
Molecular Formula: C10H9FO3
Molecular Weight: 196.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 91
Flash Point (°C): 110
Specific Gravity (20/20): 1.24


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: (4-Fluorobenzoyl)acetic Acid Methyl Ester, Methyl 3-(4-Fluorophenyl)-3-oxopropionate, 3-(4-Fluorophenyl)-3-oxopropionic Acid Methyl Ester

Formula: C₁₀H₉FO₃
MW: 196.18 g/mol
Boiling Pt: 91 °C
Density: 1.24 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00000355
CAS Number: 63131-29-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b8c93adc00a382d43b6e6ee6067a80172ae514e&vid=6b8c93adc00a382d43b6e6ee6067a80172ae514e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR