2-Fluoro-4-hydroxybenzonitrile ≥98.0%

Supplier: TCI America

F0462-5G F0462-25G
TCF0462-5GEA 34.76 CAD
TCF0462-5G TCF0462-25G
2-Fluoro-4-hydroxybenzonitrile ≥98.0%
2-Fluoro-4-hydroxybenzonitrile

CAS Number: 82380-18-5
MDL Number: MFCD00192177
Molecular Formula: C7H4FNO
Molecular Weight: 137.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 125


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Cyano-3-fluorophenol, 4-Hydroxy-2-fluorobenzonitrile

Formula: C₇H₄FNO
MW: 137.11
Boiling Pt: 261
Melting Pt: 124
Density: 1.45
MDL Number: MFCD00192177
CAS Number: 82380-18-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8db13c73009b5afd247e76f8b50e71b811d0f938&vid=8db13c73009b5afd247e76f8b50e71b811d0f938&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR