N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-serine hydrate ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: N-Fmoc-L-serine, Fmoc-Ser-OH

F0454-1G F0454-5G
TCF0454-1GEA 42.66 CAD
TCF0454-1G TCF0454-5G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-serine hydrate ≥97.0% (by HPLC, titration analysis)
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-serine hydrate

CAS Number: 73724-45-5
MDL Number: MFCD04113634
Molecular Formula: C18H17NO5
Molecular Weight: 327.34
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -12.5 deg (C=1, DMF)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Fmoc-L-serine, Fmoc-Ser-OH

Formula: C₁₈H₁₇NO₅
MDL Number: MFCD04113634
CAS Number: 73724-45-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b1ac80311e84df1d89bbba3fae0703cc6457452&vid=e5898ec04ba2cbe50228119c8a3c20ad92e7c2a1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR