2'-Fluoropropiophenone ≥98.0%

Supplier: TCI America

Synonyms: o-Fluoropropiophenone

F0450-25G F0450-5G
TCF0450-25GEA 131.14 CAD
TCF0450-25G TCF0450-5G
2'-Fluoropropiophenone ≥98.0%
2'-Fluoropropiophenone

CAS Number: 446-22-0
MDL Number: MFCD00009893
Molecular Formula: C9H9FO
Molecular Weight: 152.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 79
Specific Gravity (20/20): 1.11


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Ethyl 2-Fluorophenyl Ketone

Formula: C₉H₉FO
MW: 152.17 g/mol
Density: 1.11 (20 °C)
Flash Pt: 79 °C
MDL Number: MFCD00009893
CAS Number: 446-22-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a43219e4570625b4422e358c4048054bc38e19b3&vid=a43219e4570625b4422e358c4048054bc38e19b3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR