4-Formylphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 4-Boronobenzaldehyde, 4-Formylbenzeneboronic acid, (4-Formylphenyl)metaboric acid, Dihydroxy(4-formylphenyl)borane

F0446-1G F0446-5G
TCF0446-1GEA 44.24 CAD
TCF0446-1G TCF0446-5G
4-Formylphenylboronic acid (contains varying amounts of Anhydride)
4-Formylphenylboronic acid

CAS Number: 87199-17-5
MDL Number: MFCD00151823
Molecular Formula: C7H7BO3
Molecular Weight: 149.94
Form: Crystal
Color: White
Melting point (°C): 242


Other Product Names / Synonyms: 4-Formylbenzeneboronic Acid

Formula: C₇H₇BO₃
MW: 149.94 g/mol
Melting Pt: 242 °C
MDL Number: MFCD00151823
CAS Number: 87199-17-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ea94e9f9686e508623cf0d47dec670dc9e32f93&vid=1ea94e9f9686e508623cf0d47dec670dc9e32f93&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR