3-Formylphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

F0445-25G F0445-1G F0445-5G
TCF0445-25GEA 213.3 CAD
TCF0445-25G TCF0445-1G TCF0445-5G
3-Formylphenylboronic acid (contains varying amounts of Anhydride)
3-Formylphenylboronic acid

CAS Number: 87199-16-4
MDL Number: MFCD00161356
Molecular Formula: C7H7BO3
Molecular Weight: 149.94
Form: Crystal
Color: White
Melting point (°C): 113


Other Product Names / Synonyms: 3-Formylbenzeneboronic Acid

Formula: C₇H₇BO₃
Melting Pt: 113 °C
MDL Number: MFCD00161356
CAS Number: 87199-16-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9b22424d543a4a0210e5fd8be932cf8723611e0&vid=a9b22424d543a4a0210e5fd8be932cf8723611e0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR