6-Fluoroveratraldehyde ≥98.0%
Supplier: TCI America
F0441-5G
F0441-1G
TCF0441-5GEA
173.48
CAD
TCF0441-5G
TCF0441-1G
6-Fluoroveratraldehyde ≥98.0%
6-Fluoroveratraldehyde
CAS Number: 71924-62-4
MDL Number: MFCD00061108
Molecular Formula: C9H9FO3
Molecular Weight: 184.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 98
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 6-Fluoroveratraldehyde
Formula:
C₉H₉FO₃ Melting Pt: 98 °C |
MDL Number:
MFCD00061108 CAS Number: 71924-62-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...