Furan-3-boronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

F0438-1G F0438-5G
TCF0438-1GEA 114.27 CAD
TCF0438-1G TCF0438-5G
Furan-3-boronic acid (contains varying amounts of Anhydride)
Furan-3-boronic acid

CAS Number: 55552-70-0
MDL Number: MFCD01319007
Molecular Formula: C4H5BO3
Molecular Weight: 111.89
Form: Crystal
Color: White


Other Product Names / Synonyms: Furan-3-boronic Acid, 3-Furanylboronic Acid

Formula: C₄H₅BO₃
MDL Number: MFCD01319007
CAS Number: 55552-70-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=443693a2148d1c8fb59c32f90af0793e394a2096&vid=443693a2148d1c8fb59c32f90af0793e394a2096&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR