4-Fluoro-2-methoxyphenol ≥97.0%

Supplier: TCI America

F0436-5G F0436-1G
TCF0436-5GEA 134.3 CAD
TCF0436-5G TCF0436-1G
4-Fluoro-2-methoxyphenol ≥97.0%
4-Fluoro-2-methoxyphenol

CAS Number: 450-93-1
MDL Number: MFCD00070797
Molecular Formula: C7H7FO2
Molecular Weight: 142.13
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 195
Flash Point (°C): 101
Specific Gravity (20/20): 1.26


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Fluoroguaiacol

Formula: C₇H₇FO₂
MW: 142.13 g/mol
Boiling Pt: 195 °C
Density: 1.26 (20 °C)
Flash Pt: 101 °C
MDL Number: MFCD00070797
CAS Number: 450-93-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e2136ca9411eb18a818999ce3b8e673a666d7ed&vid=8e2136ca9411eb18a818999ce3b8e673a666d7ed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR