4-Fluoro-α-methylbenzyl alcohol ≥97.0%

Supplier: TCI America

F0432-25G F0432-5G
TCF0432-25GEA 687.3 CAD
TCF0432-25G TCF0432-5G
4-Fluoro-α-methylbenzyl alcohol ≥97.0%
4-Fluoro-α-methylbenzyl alcohol

CAS Number: 403-41-8
MDL Number: MFCD00004515
Molecular Formula: C8H9FO
Molecular Weight: 140.16
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 123
Flash Point (°C): 91
Specific Gravity (20/20): 1.12


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-(4-Fluorophenyl)ethanol, 1-(4-Fluorophenyl)ethyl Alcohol

Formula: C₈H₉FO
Boiling Pt: 123 °C
Density: 1.12 (20 °C)
Flash Pt: 91 °C
MDL Number: MFCD00004515
CAS Number: 403-41-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3108d02644106d30b9f31d1c1010bcef1e16c946&vid=3108d02644106d30b9f31d1c1010bcef1e16c946&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR