5-Fluoro-o-anisaldehyde ≥97.0%

Supplier: TCI America

Synonyms: 5-Fluoro-2-anisaldehyde, 5-Fluoro-2-methoxybenzaldehyde

F0428-1G F0428-5G
TCF0428-1GEA 52.93 CAD
TCF0428-1G TCF0428-5G
5-Fluoro-o-anisaldehyde ≥97.0%
5-Fluoro-o-anisaldehyde

CAS Number: 19415-51-1
MDL Number: MFCD00143458
Molecular Formula: C8H7FO2
Molecular Weight: 154.14
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 118
Melting point (°C): 48


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 5-Fluoro-2-methoxybenzaldehyde

Formula: C₈H₇FO₂
Boiling Pt: 118 °C
Melting Pt: 48 °C
MDL Number: MFCD00143458
CAS Number: 19415-51-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2d2d82c333f537cdb2f690b5a616ca2b996d9ec&vid=d2d2d82c333f537cdb2f690b5a616ca2b996d9ec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR