6-Fluoro-m-toluic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

F0426-25G F0426-5G F0426-1G
TCF0426-25GEA 461.36 CAD
TCF0426-25G TCF0426-5G TCF0426-1G
6-Fluoro-m-toluic acid ≥98.0% (by titrimetric analysis)
6-Fluoro-m-toluic acid

CAS Number: 321-12-0
MDL Number: MFCD00092819
Molecular Formula: C8H7FO2
Molecular Weight: 154.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 160


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 6-Fluoro-m-toluic Acid

Formula: C₈H₇FO₂
Melting Pt: 160 °C
MDL Number: MFCD00092819
CAS Number: 321-12-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1570ea6bd7247d91846d46f02705fa64d5cce90&vid=d1570ea6bd7247d91846d46f02705fa64d5cce90&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR