3-Fluorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 3-Fluorobenzeneboronic acid, (3-Fluorophenyl)boronic acid, Dihydroxy(3-fluorophenyl)borane, (3-Fluorophenyl)metaboric acid

F0404-25G F0404-5G
TCF0404-25GEA 428.18 CAD
TCF0404-25G TCF0404-5G
3-Fluorophenylboronic acid (contains varying amounts of Anhydride)
3-Fluorophenylboronic acid

CAS Number: 768-35-4
MDL Number: MFCD00236042
Molecular Formula: C6H6BFO2
Molecular Weight: 139.92
Form: Crystal
Color: White


Other Product Names / Synonyms: 3-Fluorobenzeneboronic Acid

Formula: C₆H₆BFO₂
MW: 139.92 g/mol
MDL Number: MFCD00236042
CAS Number: 768-35-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96c3d517cd33a6a822102d3f0475411b279c4380&vid=96c3d517cd33a6a822102d3f0475411b279c4380&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR