Farnesyl acetate (mixture of isomers) ≥95.0%

Supplier: TCI America

F0347-25ML F0347-5ML
TCF0347-025MLEA 94.8 CAD
TCF0347-025ML TCF0347-5ML
Farnesyl acetate (mixture of isomers) ≥95.0%
Farnesyl acetate (mixture of isomers)

CAS Number: 29548-30-9
MDL Number: MFCD00036516
Molecular Formula: C17H28O2
Molecular Weight: 264.41
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 140
Flash Point (°C): 95
Specific Gravity (20/20): 0.91


Product Categories: Bioscience, Sesquiterpenes
Other Product Names / Synonyms: 3,7,11-Trimethyl-2,6,10-dodecatrien-1-ol Acetate

Formula: C₁₇H₂₈O₂
Boiling Pt: 140 °C
Density: 0.91 (20 °C)
Flash Pt: 95 °C
MDL Number: MFCD00036516
CAS Number: 29548-30-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cec28c323715378a9b7c6c794e0295a7add9f73c&vid=cec28c323715378a9b7c6c794e0295a7add9f73c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR