N-Fluorobenzenesulfonimide ≥97.0% (by titrimetric analysis)
Supplier: TCI America
F0335-5G
F0335-25G
TCF0335-005GEA
120.71
CAD
TCF0335-005G
TCF0335-025G
N-Fluorobenzenesulfonimide ≥97.0% (by titrimetric analysis)
N-Fluorobenzenesulphonimide
[Fluorinating Reagent]
CAS Number: 133745-75-2
MDL Number: MFCD00144885
Molecular Formula: C12H10FNO4S2
Molecular Weight: 315.33
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 115
Product Categories: Synthetic Organic Chemistry, Fluorinating Agents
Other Product Names / Synonyms: N-Fluorobis(phenylsulfonyl)amine, NFSI, NFBS
Formula:
C₁₂H₁₀FNO₄S₂ MW: 315.34 g/mol Melting Pt: 115 °C |
MDL Number:
MFCD00144885 CAS Number: 133745-75-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...