(Formylmethyl)triphenylphosphonium chloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
F0331-25G
F0331-5G
TCF0331-025GEA
155.11
CAD
TCF0331-025G
76220-066
(Formylmethyl)triphenylphosphonium chloride ≥98.0% (by titrimetric analysis)
(Formylmethyl)triphenylphosphonium chloride
CAS Number: 62942-43-2
MDL Number: MFCD00012003
Molecular Formula: C20H18ClOP
Molecular Weight: 340.79
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Formula:
C₂₀H₁₈ClOP |
MDL Number:
MFCD00012003 CAS Number: 62942-43-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...