N-Fmoc-L-valine ≥98.0%
Supplier: TCI America
F0299-5G
F0299-25G
TCF0299-005GEA
90.69
CAD
TCF0299-005G
TCF0299-25G
N-Fmoc-L-valine ≥98.0%
N-Fmoc-L-valine
CAS Number: 68858-20-8
MDL Number: MFCD00037124
Molecular Formula: C20H21NO4
Molecular Weight: 339.39
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 145
Specific rotation [a]20/D: -17.5 deg (C=1, DMF)
Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Fmoc-L-valine, Fmoc-Val-OH
Formula:
C₂₀H₂₁NO₄ MW: 339.4 g/mol Melting Pt: 145 °C |
MDL Number:
MFCD00037124 CAS Number: 68858-20-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...