Fmoc-Pro-OH ≥98.0%

Supplier: TCI America

Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-proline, N-Fmoc-L-proline

F0298-25G F0298-5G
TCF0298-25GEA 510.34 CAD
TCF0298-25G TCF0298-005G
Fmoc-Pro-OH ≥98.0%
Fmoc-Pro-OH

CAS Number: 71989-31-6
MDL Number: MFCD00037122
Molecular Formula: C20H19NO4
Molecular Weight: 337.38
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 115
Specific rotation [a]20/D: -32 deg (C=1, DMF)


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Fmoc-L-proline, Fmoc-Pro-OH

Formula: C₂₀H₁₉NO₄
MW: 337.38 g/mol
Melting Pt: 115 °C
MDL Number: MFCD00037122
CAS Number: 71989-31-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7bfc002595792bedbe4d3ae3b9a68758f562076c&vid=7bfc002595792bedbe4d3ae3b9a68758f562076c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR