Fmoc-Met-OH ≥98.0%

Supplier: TCI America

Synonyms: N-Fmoc-L-methionine, FMOC-L-Methionine

F0296-25G F0296-5G
TCF0296-25GEA 97.96 CAD
TCF0296-25G TCF0296-001G TCF0296-005G
Fmoc-Met-OH ≥98.0%
Fmoc-Met-OH

CAS Number: 71989-28-1
MDL Number: MFCD00037134
Molecular Formula: C20H21NO4S
Molecular Weight: 371.45
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 141
Specific rotation [a]20/D: -29.5 deg (C=1, DMF)


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Fmoc-L-methionine, Fmoc-Met-OH

Formula: C₂₀H₂₁NO₄S
MW: 371.46 g/mol
Melting Pt: 141 °C
MDL Number: MFCD00037134
CAS Number: 71989-28-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f861458d923925123c25ff8d3a6fff6940950be5&vid=f861458d923925123c25ff8d3a6fff6940950be5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR