N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-isoleucine
Supplier: TCI America
F0294-25G
F0294-5G
TCF0294-25GEA
214.88
CAD
TCF0294-25G
TCF0294-005G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-isoleucine
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-isoleucine
CAS Number: 71989-23-6
MDL Number: MFCD00037125
Molecular Formula: C21H23NO4
Molecular Weight: 353.42
Form: Crystal
Color: White
Melting point (°C): 146
Specific rotation [a]20/D: -12 deg (C=1, DMF)
Other Product Names / Synonyms: N-Fmoc-L-isoleucine, Fmoc-Ile-OH
Formula:
C₂₁H₂₃NO₄ Melting Pt: 146 °C |
MDL Number:
MFCD00037125 CAS Number: 71989-23-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...